Learn More
2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
$128.87 - $128.87
Chemical Identifiers
CAS | 156-28-5 |
---|---|
Molecular Formula | C8H11N·ClH |
Molecular Weight (g/mol) | 157.64 |
InChI Key | SKHIBNDAFWIOPB-UHFFFAOYSA-N |
Synonym | 2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d |
PubChem CID | 9075 |
IUPAC Name | 2-phenylethanamine;hydrochloride |
SMILES | C1=CC=C(C=C1)CCN.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC441910250
|
Thermo Scientific Chemicals
441910250 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
156-28-5 | |
157.64 | |
2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d | |
2-phenylethanamine;hydrochloride |
C8H11N·ClH | |
SKHIBNDAFWIOPB-UHFFFAOYSA-N | |
9075 | |
C1=CC=C(C=C1)CCN.Cl |
Specifications
156-28-5 | |
100.0 | |
White | |
97.5 to max. 102.5% min. (Argentometry) | |
C8H11N·ClH | |
15,7339 | |
SKHIBNDAFWIOPB-UHFFFAOYSA-N | |
2-phenylethanamine;hydrochloride | |
9075 | |
98% | |
2-Phenylethylamine hydrochloride |
97.5 | |
220°C to 223°C | |
Authentic | |
Glass Bottle | |
25 g | |
2-phenylethylamine hydrochloride, 2-phenylethanamine hydrochloride, phenethylamine hydrochloride, benzeneethanamine, hydrochloride, phenethylammonium chloride, beta-phenylethylamine hydrochloride, usaf el-76, 1-phenyl-2-aminoethane hydrochloride, beta-phenethylamine hydrochloride, unii-vof61xw69d | |
C1=CC=C(C=C1)CCN.Cl | |
157.64 | |
157.64 | |
Crystalline Powder or Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 205-849-2
RUO – Research Use Only