missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Phenylethylamine Hydroiodide 98.0+%, TCI America™

Supplier:  TCI America P22135G

 View more versions of this product

Catalog No. P22135G


Explore available promotions

Description

Description

Specifications

Specifications

2-Phenylethylamine Hydroiodide
259°C
C8H12IN
Phenethylamine Hydroiodide, 2-Phenylethylammonium Iodide, Phenethylammonium Iodide
C1=CC=C(C=C1)CCN.I
249.095
249.10
Crystalline Powder
151059-43-7
White
5 g
UPHCENSIMPJEIS-UHFFFAOYSA-N
2-phenylethanamine;hydroiodide
91972166
≥98.0% (HPLC,N)

Chemical Identifiers

151059-43-7
249.095
Phenethylamine Hydroiodide, 2-Phenylethylammonium Iodide, Phenethylammonium Iodide
2-phenylethanamine;hydroiodide
C8H12IN
UPHCENSIMPJEIS-UHFFFAOYSA-N
91972166
C1=CC=C(C=C1)CCN.I
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions