missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylquinoline, 99+%
CAS: 612-96-4 | C15H11N | 205.26 g/mol
$129.59 - $129.59
Identifiants chimiques
| CAS | 612-96-4 |
|---|---|
| Molecular Formula | C15H11N |
| Molecular Weight (g/mol) | 205.26 |
| MDL Number | MFCD00011568 |
| InChI Key | FSEXLNMNADBYJU-UHFFFAOYSA-N |
| Synonym | quinoline, 2-phenyl, 2-phenyl quinoline, .alpha.-phenylquinoline, 2-phenyl-quinoline, quinoline, phenyl, chembl89786, a-phenylquinoline, quinoline,phenyl, phenylquinoline, pubchem15940 |
| PubChem CID | 71545 |
| IUPAC Name | 2-phenylquinoline |
| SMILES | C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AAH3192003
|
Thermo Scientific Chemicals
H3192003 |
1 g |
chaque for $129.59
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 612-96-4 | |
| 205.26 | |
| FSEXLNMNADBYJU-UHFFFAOYSA-N | |
| 71545 | |
| C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1 |
| C15H11N | |
| MFCD00011568 | |
| quinoline, 2-phenyl, 2-phenyl quinoline, .alpha.-phenylquinoline, 2-phenyl-quinoline, quinoline, phenyl, chembl89786, a-phenylquinoline, quinoline,phenyl, phenylquinoline, pubchem15940 | |
| 2-phenylquinoline |
Spécifications
| 612-96-4 | |
| 363°C | |
| MFCD00011568 | |
| quinoline, 2-phenyl, 2-phenyl quinoline, .alpha.-phenylquinoline, 2-phenyl-quinoline, quinoline, phenyl, chembl89786, a-phenylquinoline, quinoline,phenyl, phenylquinoline, pubchem15940 | |
| C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1 | |
| 205.26 | |
| 205.26 | |
| 2-Phenylquinoline |
| 82°C to 84°C | |
| C15H11N | |
| 1 g | |
| FSEXLNMNADBYJU-UHFFFAOYSA-N | |
| 2-phenylquinoline | |
| 71545 | |
| ≥99% |
Sécurité et manipulation
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-326-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only