Learn More
2-Picolyl chloride hydrochloride, 98%, Thermo Scientific Chemicals
$212.76 - $212.76
Chemical Identifiers
CAS | 6959-47-3 |
---|---|
Molecular Formula | C6H6ClN·HCl |
Molecular Weight (g/mol) | 164.04 |
MDL Number | MFCD00012811 |
InChI Key | JPMRGPPMXHGKRO-UHFFFAOYSA-N |
Synonym | 2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0 |
PubChem CID | 23392 |
ChEBI | CHEBI:76600 |
IUPAC Name | 2-(chloromethyl)pyridine;hydrochloride |
SMILES | C1=CC=NC(=C1)CCl.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC172120250
|
Thermo Scientific Chemicals
172120250 |
25 g | Glass bottle |
Each for $212.76
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6959-47-3 | |
164.04 | |
JPMRGPPMXHGKRO-UHFFFAOYSA-N | |
23392 | |
2-(chloromethyl)pyridine;hydrochloride |
C6H6ClN·HCl | |
MFCD00012811 | |
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0 | |
CHEBI:76600 | |
C1=CC=NC(=C1)CCl.Cl |
Specifications
6959-47-3 | |
100.0 | |
Brown to Green-Yellow | |
98% | |
C6H6ClN·HCl | |
25 g | |
JPMRGPPMXHGKRO-UHFFFAOYSA-N | |
2-(chloromethyl)pyridine;hydrochloride | |
23392 | |
164.04 | |
Crystalline Mass |
97.5 | |
119.0°C to 125.0°C | |
Authentic | |
Glass bottle | |
MFCD00012811 | |
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0 | |
C1=CC=NC(=C1)CCl.Cl | |
164.04 | |
CHEBI:76600 | |
98% | |
2-Picolyl chloride hydrochloride, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 230-149-9
RTECSNumber : US6825000
TSCA : TSCA
RUO – Research Use Only