Learn More
2-Propanethiol, 98%, Thermo Scientific Chemicals
$38.19 - $115.30
Chemical Identifiers
CAS | 75-33-2 |
---|---|
Molecular Formula | C3H8S |
Molecular Weight (g/mol) | 76.157 |
MDL Number | MFCD00004863 |
InChI Key | KJRCEJOSASVSRA-UHFFFAOYSA-N |
Synonym | 2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol |
PubChem CID | 6364 |
ChEBI | CHEBI:8474 |
IUPAC Name | propane-2-thiol |
SMILES | CC(C)S |
Description
2-Propanethiol Reacts with 3-chloro-propene to get 3-isopropylsulfanyl-propene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
75-33-2 | |
76.157 | |
KJRCEJOSASVSRA-UHFFFAOYSA-N | |
6364 | |
propane-2-thiol |
C3H8S | |
MFCD00004863 | |
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol | |
CHEBI:8474 | |
CC(C)S |
Specifications
75-33-2 | |
Colorless | |
57°C to 60°C | |
Stench | |
1.4255 | |
MFCD00004863 | |
UN2402 | |
Air Sensitive | |
Slightly soluble in water. | |
CC(C)S | |
76.157 | |
CHEBI:8474 | |
98% | |
2-Propanethiol |
-131°C | |
0.82 g/mL | |
-34° (-29°F) | |
C3H8S | |
(CH3)2CHSH | |
25 mL | |
605260 | |
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol | |
KJRCEJOSASVSRA-UHFFFAOYSA-N | |
propane-2-thiol | |
6364 | |
76.16 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P261-P272-P280-P303+P361+P353-P333+P313-P363-P370+P378q-P501c
H225-H317
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPANETHIOLS
EINECSNumber : 200-861-4
RTECSNumber : TZ7302000
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only