Learn More
2-Propanol, ACS Grade, Fisher Chemical™
In addition to meeting or exceeding the latest ACS specifications, are analyzed for more than 16 metals; Actual lot of analysis on label
$179.69 - $1,845.18
Chemical Identifiers
| CAS | 67-63-0 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.096 |
| MDL Number | MFCD00011674 |
| InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
| Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
| PubChem CID | 3776 |
| ChEBI | CHEBI:17824 |
| IUPAC Name | propan-2-ol |
| SMILES | CC(C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
A416-1
|
Thermo Fisher Scientific
BPA4161 |
1 L | HDPE Bottle |
N/A
|
|
||||
|
A416-4
|
Fisher Chemical
BPA4164 |
4 L | Glass Bottle |
N/A
|
|
||||
|
A416P-4
|
Thermo Fisher Scientific
BPA416P4 |
4 L | Poly Bottle |
N/A
|
|
||||
|
A416-20
|
Fisher Chemical
BPA41620 |
20 L | Steel Pail |
N/A
|
|
||||
|
A416-200
|
Thermo Fisher Scientific
BPA416200 |
200 L | Steel Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 67-63-0 | |
| 60.096 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| 3776 | |
| propan-2-ol |
| C3H8O | |
| MFCD00011674 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CHEBI:17824 | |
| CC(C)O |
Specifications
| 7 | |
| 67-63-0 | |
| 0.785g/cm³ | |
| 1 L | |
| Pass Test | |
| Liquid | |
| 0.001% max. | |
| 0.785 | |
| (CH3)2CHOH | |
| 43mmHg at 20°C | |
| 1219 | |
| isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
| CC(C)O | |
| 60.096 | |
| CHEBI:17824 | |
| ≥99.5% | |
| 2-Propanol |
| -89.5°C | |
| Colorless | |
| 81°C | |
| 12°C (53.6°F) | |
| HDPE Bottle | |
| 1.3740 to 1.3760 (at 25°C) | |
| ≥99.5 % | |
| C3H8O | |
| MFCD00011674 | |
| 2.27 mPa/s at 20°C | |
| 0.2% max. | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 3776 | |
| 60.1 | |
| Certified ACS Reagent |
Safety and Handling
WARNING!
Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid