missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanol For spectroscopy Uvasol™, MilliporeSigma™

For spectroscopy Uvasol™

Supplier:  Technidata 1009931000

 View more versions of this product

Catalog No. M1009931000


Explore available promotions

Description

Description

Specifications

Specifications

Neutral (at 20°C)
-89.5°C
Colorless
82.4°C
12°C (54°F)
Alcohol-like
Liquid
C3H8O
1219
Soluble in water at 20°C
CC(C)O
60.096
CHEBI:17824
2-Propanol
67-63-0
0.78g/cm3 at 20°C
1 L
425°C (797.9°F)
Glass Bottle
1.37
43 hPa (20°C)
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1

Chemical Identifiers

67-63-0
60.096
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

H336: May cause drowsiness or dizziness.

H225: Highly flammable liquid and vapour.

Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.

SDS
Documents

Documents

Promotions

Promotions