missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Propanol for HPLC, 99.5%, MilliporeSigma™ Supelco™
$1505.61 - $1505.61
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.10 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
111005327
|
Merck Emd Millipore
43920720LP24B |
20 L | PVC-coated Bottle |
Each for $1,505.61
|
|
||||
Description
2-Propanol (i-PrOH), a renewable secondary alcohol, is water soluble, colorless, volatile liquid with a sweet odor. It is a potential substitute to petroleum-derived gasoline. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range, 277-346K. Its photocatalytic degradation has been examined by various methods.Chemical Identifiers
67-63-0 | |
60.10 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol | |
CC(C)O |
Specifications
-89.5°C | |
0.785 g/mL (at 25°C) | |
20 L | |
n20/D 1.377 | |
(CH3)2 CHOH | |
44 mmHg (25°C) | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
60.10 | |
HPLC |
67-63-0 | |
82°C | |
PVC-coated Bottle | |
C3H8O | |
MFCD00011674 | |
sec-Propyl alcohol; IPA; Isopropanol; Isopropyl alcohol | |
CC(C)O | |
60.10 | |
99.5% |
Safety and Handling
RTECSNumber : NT8050000