missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Propanol (HPLC), Fisher Chemical™

HPLC mobile phase for analytical and preparative separations

Supplier:  Thermo Fisher Scientific BPA4514

 View more versions of this product

Catalog No. A451-4



Explore available promotions

Description

Description

Specifications

Specifications

7
-89.5°C
0.025 AU max. at 254nm, 0.13 AU max. at 230nm, 0.25 AU max. at 220nm, 1.00 AU max. at 205nm
0.785g/cm³
4 L
Glass Bottle
Mobile phase for HPLC applications
2ppm max.
C3H8O
43mmHg at 20°C
0.10% max.
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
HPLC
2-Propanol
67-63-0
Colorless
81°C
Pass Test
Liquid
1.3740 to 1.3760 (at 25°C)
99.9% min by GC
MFCD00011674
2.27 mPa/s at 20°C
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
≥99.9%

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions

Safe-Cote™ bottles that provide the purity of glass and most of the benefits for storing and dispensing chemicals. If they break, glass fragments and liquids are more likely to remain trapped.