missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Propanol, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS
Supplier: Honeywell-Chromasolv 349656X1L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
2-Propanol | |
CHROMASOLV™ LC-MS | |
100.0000% | |
77°C to 83°C | |
12°C | |
C3H8O | |
MFCD00011674 | |
635639 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.11 | |
LC-MS |
-89.5°C (literature) | |
67-63-0 | |
0.785Â g/mLÂ at 25Â °C(lit.) | |
6 x 1 L | |
Glass Bottle | |
(CH3)2CHOH | |
UN1219 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
>99.9% |
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Safety and Handling
ShelfLife : 360 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Isopropanol
EINECSNumber : 200-661-7
Recommended Storage : Room Temp