missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyrazinylmethanol, 97%, Thermo Scientific™

$299.45 - $299.45

Chemical Identifiers

CAS 6705-33-5
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD01693938
InChI Key LFCWHDGQCWJKCG-UHFFFAOYSA-N
Synonym 2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol
PubChem CID 201734
SMILES OCC1=CN=CC=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC03409DA
SDS View Documents
Thermo Scientific Chemicals
CC03409DA
1 g Amber Glass Bottle N/A
 
Description

Description

Specifications

Chemical Identifiers

6705-33-5
110.12
LFCWHDGQCWJKCG-UHFFFAOYSA-N
201734
C5H6N2O
MFCD01693938
2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol
OCC1=CN=CC=N1

Specifications

6705-33-5
100.0
Amber Glass Bottle
MFCD01693938
2-pyrazinylmethanol, pyrazin-2-yl methanol, pyrazinemethanol, 2-hydroxymethyl pyrazine, 2-pyrazinemethanol, 2-hydroxymethylpyrazine, pyrazin-2-yl-methanol, unii-c27z7qi77d, pyrazin-2-ylmethan-1-ol
OCC1=CN=CC=N1
110.12
110.12
2-Pyrazinylmethanol
94.5
95%
C5H6N2O
1 g
LFCWHDGQCWJKCG-UHFFFAOYSA-N
pyrazin-2-ylmethanol
201734
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'