missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Pyridylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H58544AD

Catalog No. AAH58544AD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of P,N ligands for asymmetric Ir-catalyzed reactions.

Solubility
Reacts with water.

Notes
Handle under dry protective gas. Air sensitive. Store away from strong bases a
Specifications

Specifications

2-Pyridylzinc bromide
218777-23-2
C5H5BrNZn-
50 mL
Air sensitive; May form a precipitate
Reacts with water.
C1=CC=NC=C1.[Zn].[Br-]
224.386
223.38
Packaged under Argon in resealable ChemSeal™ bottles
0.974
MFCD00671985
UN3399
2-pyridylzinc bromide
WUTSMSLAOBVVCP-UHFFFAOYSA-M
pyridine;zinc;bromide
76056926

Chemical Identifiers

218777-23-2
224.386
WUTSMSLAOBVVCP-UHFFFAOYSA-M
76056926
C1=CC=NC=C1.[Zn].[Br-]
C5H5BrNZn-
MFCD00671985
2-pyridylzinc bromide
pyridine;zinc;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335-H336-H351

DOTInformation : Hazard Class: 3; Packaging Group: II

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only