missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Sulfobenzoic acid hydrate, 98%, Thermo Scientific Chemicals

$76.70 - $208.18

Chemical Identifiers

CAS 123333-68-6
Molecular Formula C7H6O5S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00007479
InChI Key ZMPRRFPMMJQXPP-UHFFFAOYSA-N
Synonym 2-sulfobenzoic acid hydrate, benzoic acid, 2-sulfo-hydrate, acmc-20anmu, 2-sulphobenzoic acid hydrate, c7h6o5s.h2o, 2-sulfobenzoic acid, oxamethane, benzoic acid, 2-sulfo-, monohydrate
PubChem CID 24820382
IUPAC Name 2-sulfobenzoic acid;hydrate
SMILES OC(=O)C1=CC=CC=C1S(O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2338114
View Documents
Thermo Scientific Chemicals
B2338114
25 g
Each for $76.70
 
AAB2338122
View Documents
Thermo Scientific Chemicals
B2338122
100 g
Each for $208.18
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123333-68-6
202.18
ZMPRRFPMMJQXPP-UHFFFAOYSA-N
24820382
OC(=O)C1=CC=CC=C1S(O)(=O)=O
C7H6O5S
MFCD00007479
2-sulfobenzoic acid hydrate, benzoic acid, 2-sulfo-hydrate, acmc-20anmu, 2-sulphobenzoic acid hydrate, c7h6o5s.h2o, 2-sulfobenzoic acid, oxamethane, benzoic acid, 2-sulfo-, monohydrate
2-sulfobenzoic acid;hydrate

Specifications

123333-68-6
C7H6O5S
25 g
2106852
2-sulfobenzoic acid hydrate, benzoic acid, 2-sulfo-hydrate, acmc-20anmu, 2-sulphobenzoic acid hydrate, c7h6o5s.h2o, 2-sulfobenzoic acid, oxamethane, benzoic acid, 2-sulfo-, monohydrate
OC(=O)C1=CC=CC=C1S(O)(=O)=O
202.18
202.19 (Anhydrous)
2-Sulfobenzoic acid hydrate
∼68°C to 72°C
MFCD00007479
UN3261
Hygroscopic
ZMPRRFPMMJQXPP-UHFFFAOYSA-N
2-sulfobenzoic acid;hydrate
24820382
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 211-176-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only