missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Tetrahydropyran-4-yloxy)aniline, 95%, Thermo Scientific™

Chemical Identifiers

CAS 898289-35-5
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD09025872
InChI Key CLGPZXHLRKSEDD-UHFFFAOYSA-N
Synonym 2-tetrahydro-2h-pyran-4-yloxy aniline, 2-tetrahydropyran-4-yloxy aniline, 2-tetrahydro-2h-pyran-4-yl oxy aniline, 2-oxan-4-yloxy aniline, benzenamine,2-tetrahydro-2h-pyran-4-yl oxy, benzenamine, 2-tetrahydro-2h-pyran-4-yl oxy, 2-oxan-4-yl oxy aniline, 2-tetrahydropyran-4-yloxyaniline, 4-2-aminophenoxy tetrahydro-2h-pyran, 2-tetrahydro-2h-pyran-4-yl ;oxy ;aniline
PubChem CID 24229516
IUPAC Name 2-(oxan-4-yloxy)aniline
SMILES C1COCCC1OC2=CC=CC=C2N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC52214CB
SDS View Documents
Thermo Scientific Chemicals
CC52214CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC52214DA
SDS View Documents
Thermo Scientific Chemicals
CC52214DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

898289-35-5
193.246
CLGPZXHLRKSEDD-UHFFFAOYSA-N
24229516
C1COCCC1OC2=CC=CC=C2N
C11H15NO2
MFCD09025872
2-tetrahydro-2h-pyran-4-yloxy aniline, 2-tetrahydropyran-4-yloxy aniline, 2-tetrahydro-2h-pyran-4-yl oxy aniline, 2-oxan-4-yloxy aniline, benzenamine,2-tetrahydro-2h-pyran-4-yl oxy, benzenamine, 2-tetrahydro-2h-pyran-4-yl oxy, 2-oxan-4-yl oxy aniline, 2-tetrahydropyran-4-yloxyaniline, 4-2-aminophenoxy tetrahydro-2h-pyran, 2-tetrahydro-2h-pyran-4-yl ;oxy ;aniline
2-(oxan-4-yloxy)aniline

Specifications

898289-35-5
100.0
Amber Glass Bottle
MFCD09025872
2-tetrahydro-2h-pyran-4-yloxy aniline, 2-tetrahydropyran-4-yloxy aniline, 2-tetrahydro-2h-pyran-4-yl oxy aniline, 2-oxan-4-yloxy aniline, benzenamine,2-tetrahydro-2h-pyran-4-yl oxy, benzenamine, 2-tetrahydro-2h-pyran-4-yl oxy, 2-oxan-4-yl oxy aniline, 2-tetrahydropyran-4-yloxyaniline, 4-2-aminophenoxy tetrahydro-2h-pyran, 2-tetrahydro-2h-pyran-4-yl ;oxy ;aniline
C1COCCC1OC2=CC=CC=C2N
193.246
193.24
2-(Tetrahydropyran-4-yloxy)aniline
94.5
95%
C11H15NO2
250mg
CLGPZXHLRKSEDD-UHFFFAOYSA-N
2-(oxan-4-yloxy)aniline
24229516
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'