missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Thenylmercaptan, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 6258-63-5 |
|---|---|
| Molecular Formula | C5H6S2 |
| Molecular Weight (g/mol) | 130.22 |
| MDL Number | MFCD00107135 |
| InChI Key | GCZQHDFWKVMZOE-UHFFFAOYSA-N |
| Synonym | 2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene |
| PubChem CID | 80408 |
| IUPAC Name | thiophen-2-ylmethanethiol |
| SMILES | C1=CSC(=C1)CS |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC265110010
|
Thermo Scientific Chemicals
265110010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC265110050
|
Thermo Scientific Chemicals
265110050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 6258-63-5 | |
| 130.22 | |
| GCZQHDFWKVMZOE-UHFFFAOYSA-N | |
| 80408 | |
| C1=CSC(=C1)CS |
| C5H6S2 | |
| MFCD00107135 | |
| 2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene | |
| thiophen-2-ylmethanethiol |
Specifications
| 6258-63-5 | |
| 100.0 | |
| 90°C (15.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.6020 to 1.604 | |
| 1g | |
| 2-thiophenemethanethiol, thenyl mercaptan, 2-thenylmercaptan, 2-thienylmethanethiol, thiophene-2-methanethiol, 2-thenylthiol, thiophen-2-yl-methanethiol, unii-9jml279xs6, 2-thenyl mercaptan, 2-mercaptomethyl thiophene | |
| GCZQHDFWKVMZOE-UHFFFAOYSA-N | |
| thiophen-2-ylmethanethiol | |
| 80408 | |
| 98% |
| 97.5 | |
| 1.2000g/mL | |
| 81°C | |
| 98% | |
| C5H6S2 | |
| MFCD00107135 | |
| 1.2 | |
| Solubility in water: slightly soluble | |
| C1=CSC(=C1)CS | |
| 130.22 | |
| 130.22 | |
| 2-Thenylmercaptan |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 228-394-1
RUO â Research Use Only