Learn More
(2-Thien-2-ylphenyl)methanol, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 773872-97-2 |
|---|---|
| Molecular Formula | C11H10OS |
| Molecular Weight (g/mol) | 190.26 |
| MDL Number | MFCD06203081 |
| InChI Key | IUULMJAEIKVTKZ-UHFFFAOYSA-N |
| Synonym | 2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol |
| PubChem CID | 2795559 |
| IUPAC Name | [2-(thiophen-2-yl)phenyl]methanol |
| SMILES | OCC1=CC=CC=C1C1=CC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
CC35509CB
|
Thermo Scientific Chemicals
CC35509CB |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC35509DA
|
Thermo Scientific Chemicals
CC35509DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC35509EA
|
Thermo Scientific Chemicals
CC35509EA |
10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 773872-97-2 | |
| 190.26 | |
| IUULMJAEIKVTKZ-UHFFFAOYSA-N | |
| 2795559 | |
| OCC1=CC=CC=C1C1=CC=CS1 |
| C11H10OS | |
| MFCD06203081 | |
| 2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol | |
| [2-(thiophen-2-yl)phenyl]methanol |
Specifications
| 773872-97-2 | |
| 100.0 | |
| Amber glass bottle | |
| MFCD06203081 | |
| 2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol | |
| OCC1=CC=CC=C1C1=CC=CS1 | |
| 190.26 | |
| 190.27 | |
| (2-Thien-2-ylphenyl)methanol |
| 96.5 | |
| 97% | |
| C11H10OS | |
| 250mg | |
| IUULMJAEIKVTKZ-UHFFFAOYSA-N | |
| (2-thiophen-2-ylphenyl)methanol | |
| 2795559 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning