Learn More
2-(Tri-n-butylstannyl)oxazole, 95%, Thermo Scientific Chemicals
$130.74 - $1463.36
Chemical Identifiers
CAS | 145214-05-7 |
---|---|
Molecular Formula | C15H29NOSn |
Molecular Weight (g/mol) | 358.113 |
MDL Number | MFCD06798076 |
InChI Key | YOWGRWHKDCHINP-UHFFFAOYSA-N |
Synonym | 2-tributylstannyl oxazole, 2-tri-n-butylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1, 2-tributylstannyl-oxazole, 2-tributylstannanyloxazole |
PubChem CID | 15486183 |
IUPAC Name | tributyl(1,3-oxazol-2-yl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)C1=NC=CO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH55870MD
|
Thermo Scientific Chemicals
H55870MD |
250 mg |
Each for $130.74
|
|
||||||
AAH5587003
|
Thermo Scientific Chemicals
H5587003 |
1 g |
Each for $372.26
|
|
||||||
AAH5587006
|
Thermo Scientific Chemicals
H5587006 |
5 g |
Each for $1,463.36
|
|
||||||
Description
2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Tri-n-butylstannyl)oxazole act as a synthetic building block, which is utilized in Stille coupling
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
145214-05-7 | |
358.113 | |
YOWGRWHKDCHINP-UHFFFAOYSA-N | |
15486183 | |
CCCC[Sn](CCCC)(CCCC)C1=NC=CO1 |
C15H29NOSn | |
MFCD06798076 | |
2-tributylstannyl oxazole, 2-tri-n-butylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1, 2-tributylstannyl-oxazole, 2-tributylstannanyloxazole | |
tributyl(1,3-oxazol-2-yl)stannane |
Specifications
145214-05-7 | |
1.71 g/mL | |
>110°C (230°F) | |
1.493 | |
250 mg | |
2-tributylstannyl oxazole, 2-tri-n-butylstannyl oxazole, 2-tributylstannyl-1,3-oxazole, 2-tributylstannanyl-oxazole, 2-tributylstannyloxazole, 2tributylstannyloxazole, acmc-20aom1, 2-tributylstannyl-oxazole, 2-tributylstannanyloxazole | |
YOWGRWHKDCHINP-UHFFFAOYSA-N | |
tributyl(1,3-oxazol-2-yl)stannane | |
15486183 | |
97% | |
2-(Tri-n-butylstannyl)oxazole |
Colorless | |
108°C to 110°C (0.2 mmHg) | |
C15H29NOSn | |
MFCD06798076 | |
UN2788 | |
Insoluble in water. | |
CCCC[Sn](CCCC)(CCCC)C1=NC=CO1 | |
358.113 | |
358.11 | |
Liquid |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only