missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Trifluoromethoxyphenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 175676-65-0
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320763
InChI Key AIJCNTOYZPKURP-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
PubChem CID 2777204
IUPAC Name [2-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1OC(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC393810010
SDS View Documents
Thermo Scientific Chemicals
393810010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC393810050
SDS View Documents
Thermo Scientific Chemicals
393810050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

175676-65-0
205.93
AIJCNTOYZPKURP-UHFFFAOYSA-N
2777204
OB(O)C1=CC=CC=C1OC(F)(F)F
C7H6BF3O3
MFCD01320763
2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
[2-(trifluoromethoxy)phenyl]boronic acid

Specifications

175676-65-0
100.0
96% min. (HPLC)
C7H6BF3O3
MFCD01320763
2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid, 2-trifluormethoxyphenylboronic acid, o-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxy phenyl boranediol, boronic acid, 2-trifluoromethoxy phenyl
OB(O)C1=CC=CC=C1OC(F)(F)F
205.93
205.93
2-Trifluoromethoxyphenylboronic acid
96.0
Authentic
Glass Bottle
F3COC6H4B(OH)2
1g
AIJCNTOYZPKURP-UHFFFAOYSA-N
[2-(trifluoromethoxy)phenyl]boronic acid
2777204
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only