missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Trifluoromethyl)pyridine-5-boronic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6410606

 View more versions of this product

Catalog No. AAH6410606


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-(Trifluoromethyl)pyridine-5-boronic acid
C6H5BF3NO2
5 g
Slightly soluble in water.
B(C1=CN=C(C=C1)C(F)(F)F)(O)O
190.916
190.92
Solid
868662-36-6
MFCD07375381
2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid
BNTIPMNMTIAWIW-UHFFFAOYSA-N
[6-(trifluoromethyl)pyridin-3-yl]boronic acid
25134278
98%

Chemical Identifiers

868662-36-6
190.916
BNTIPMNMTIAWIW-UHFFFAOYSA-N
25134278
B(C1=CN=C(C=C1)C(F)(F)F)(O)O
C6H5BF3NO2
MFCD07375381
2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid
[6-(trifluoromethyl)pyridin-3-yl]boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only