Learn More
2-(Trifluoromethyl)pyridine-5-boronic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H64106MD
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2-(Trifluoromethyl)pyridine-5-boronic acid | |
C6H5BF3NO2 | |
250 mg | |
Slightly soluble in water. | |
B(C1=CN=C(C=C1)C(F)(F)F)(O)O | |
190.916 | |
190.92 | |
Solid |
868662-36-6 | |
MFCD07375381 | |
2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid | |
BNTIPMNMTIAWIW-UHFFFAOYSA-N | |
[6-(trifluoromethyl)pyridin-3-yl]boronic acid | |
25134278 | |
98% |
Chemical Identifiers
868662-36-6 | |
190.916 | |
BNTIPMNMTIAWIW-UHFFFAOYSA-N | |
25134278 | |
B(C1=CN=C(C=C1)C(F)(F)F)(O)O |
C6H5BF3NO2 | |
MFCD07375381 | |
2-trifluoromethyl pyridine-5-boronic acid, 6-trifluoromethyl pyridin-3-yl boronic acid, 2-trifluoromethyl-5-pyridineboric acid, 2-trifluoromethyl pyridin-5-ylboronic acid, 6-trifluoromethyl pyridine-3-boronic acid, 6-trifluoromethyl pyridin-3-ylboronic acid, 2-trifluoromethylpyridine-5-boronic acid, 2-trifluoromethyl-5-pyridineboronic acid, 6-trifluoromethyl-3-pyridylboronic acid | |
[6-(trifluoromethyl)pyridin-3-yl]boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only