Learn More
2-(Trimethylsilyl)thiazole, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 79265-30-8 |
---|---|
Molecular Formula | C6H11NSSi |
Molecular Weight (g/mol) | 157.31 |
MDL Number | MFCD00066274 |
InChI Key | VJCHUDDPWPQOLH-UHFFFAOYSA-N |
Synonym | 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq |
PubChem CID | 588453 |
IUPAC Name | trimethyl(1,3-thiazol-2-yl)silane |
SMILES | C[Si](C)(C)C1=NC=CS1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC292290050
|
Thermo Scientific Chemicals
292290050 |
5mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC292290250
|
Thermo Scientific Chemicals
292290250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
79265-30-8 | |
157.31 | |
VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
588453 | |
C[Si](C)(C)C1=NC=CS1 |
C6H11NSSi | |
MFCD00066274 | |
2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
trimethyl(1,3-thiazol-2-yl)silane |
Specifications
79265-30-8 | |
100.0 | |
53.0°C to 55.0°C (10.0mmHg) | |
Authentic | |
Glass bottle | |
1.4950 to 1.5000 | |
5mL | |
2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
trimethyl(1,3-thiazol-2-yl)silane | |
588453 | |
96% |
95.0 | |
0.9800g/mL | |
62°C | |
95% min. (GC) | |
C6H11NSSi | |
MFCD00066274 | |
0.98 | |
Solubility in water: insoluble | |
C[Si](C)(C)C1=NC=CS1 | |
157.31 | |
157.31 | |
2-(Trimethylsilyl)thiazole |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO â Research Use Only