missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylpyridine (stabilized with TBC) 97.0+%, TCI America™

$44.73 - $303.84

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006355
InChI Key KGIGUEBEKRSTEW-UHFFFAOYSA-N
Synonym 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
PubChem CID 7521
IUPAC Name 2-ethenylpyridine
SMILES C=CC1=CC=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
V002425ML
View Documents
TCI America
V002425ML
25 mL
Each for $44.73
Only null left
 
V0024500ML
View Documents
TCI America
V0024500ML
500 mL
Each for $303.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

100-69-6
105.14
KGIGUEBEKRSTEW-UHFFFAOYSA-N
7521
C=CC1=CC=CC=N1
C7H7N
MFCD00006355
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
2-ethenylpyridine

Specifications

100-69-6
158°C
MFCD00006355
3073
KGIGUEBEKRSTEW-UHFFFAOYSA-N
2-ethenylpyridine
7521
≥97.0% (GC)
2-Vinylpyridine (stabilized with TBC)
-50°C
C7H7N
25 mL
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
C=CC1=CC=CC=N1
105.14
105.14
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0716

RTECSNumber : UU1040000

TSCA : Yes

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'