missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2H-1,4-Benzoxazin-3(4H)-one, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2762214

 View more versions of this product

Catalog No. AAH2762214


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2H-1,4-Benzoxazin-3(4H)-one
173°C to 175°C
MFCD00158536
2h-1,4-benzoxazin-3 4h-one, 2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3-one, 3-oxo-4h-benzo 1,4 oxazine, 3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3-ol, 2h-1,4-benzoxazin-3 4h-one 8ci 9ci, 4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazine-3 4h-one, 2h,4h-benzo e 1,4-oxazaperhydroin-3-one
C1C(=O)NC2=CC=CC=C2O1
149.149
149.15
5466-88-6
C8H7NO2
25 g
QRCGFTXRXYMJOS-UHFFFAOYSA-N
4H-1,4-benzoxazin-3-one
72757
99%

Chemical Identifiers

5466-88-6
149.149
QRCGFTXRXYMJOS-UHFFFAOYSA-N
72757
C1C(=O)NC2=CC=CC=C2O1
C8H7NO2
MFCD00158536
2h-1,4-benzoxazin-3 4h-one, 2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3-one, 3-oxo-4h-benzo 1,4 oxazine, 3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3-ol, 2h-1,4-benzoxazin-3 4h-one 8ci 9ci, 4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazine-3 4h-one, 2h,4h-benzo e 1,4-oxazaperhydroin-3-one
4H-1,4-benzoxazin-3-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only