missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2H-Pyran-2-one, 97%, Thermo Scientific™

Chemical Identifiers

CAS 504-31-4
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00006639
InChI Key ZPSJGADGUYYRKE-UHFFFAOYSA-N
Synonym 2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
PubChem CID 68154
ChEBI CHEBI:37965
IUPAC Name pyran-2-one
SMILES C1=CC(=O)OC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC376810050
SDS View Documents
Thermo Scientific Chemicals
376810050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

504-31-4
96.09
ZPSJGADGUYYRKE-UHFFFAOYSA-N
68154
pyran-2-one
C5H4O2
MFCD00006639
2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
CHEBI:37965
C1=CC(=O)OC=C1

Specifications

504-31-4
100.0
104°C (30.0mmHg)
Authentic
Glass bottle
1.5280 to 1.531
5g
2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
C1=CC(=O)OC=C1
96.09
CHEBI:37965
97%
96.0
1.2100g/mL
94°C
96% min. (GC)
C5H4O2
MFCD00006639
1.21
ZPSJGADGUYYRKE-UHFFFAOYSA-N
pyran-2-one
68154
96.09
2H-Pyran-2-one
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 207-990-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only