missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Thermo Scientific™

Chemical Identifiers

CAS 321744-26-7
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.30
MDL Number MFCD02259744
InChI Key RNMVWSAJMIKMDY-DTWKUNHWSA-N
Synonym 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid methyl ester, 2r,4s-1-tert-butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic aci, 2r,4s-1-tert-butyl2-methyl4-hydroxypiperidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-cis-4-hydroxy-l-pipecolinic acid methyl ester, 1-o-tert-butyl 2-o-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, o1-tert-butyl o2-methyl 2r,4s-4-oxidanylpiperidine-1,2-dicarboxylate, tert-butyl 4s,2r-4-hydroxy-2-methoxycarbonyl piperidinecarboxylate, 2r-4beta-hydroxypiperidine-1,2beta-dicarboxylic acid 1-tert-butyl 2-methyl ester
PubChem CID 2734504
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
SMILES COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC362970010
SDS View Documents
Thermo Scientific Chemicals
362970010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

321744-26-7
259.30
RNMVWSAJMIKMDY-DTWKUNHWSA-N
2734504
COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C
C12H21NO5
MFCD02259744
2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid methyl ester, 2r,4s-1-tert-butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic aci, 2r,4s-1-tert-butyl2-methyl4-hydroxypiperidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-cis-4-hydroxy-l-pipecolinic acid methyl ester, 1-o-tert-butyl 2-o-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, o1-tert-butyl o2-methyl 2r,4s-4-oxidanylpiperidine-1,2-dicarboxylate, tert-butyl 4s,2r-4-hydroxy-2-methoxycarbonyl piperidinecarboxylate, 2r-4beta-hydroxypiperidine-1,2beta-dicarboxylic acid 1-tert-butyl 2-methyl ester
1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate

Specifications

321744-26-7
100.0
Authentic
Glass bottle
MFCD02259744
2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic acid methyl ester, 2r,4s-1-tert-butyl 2-methyl 4-hydroxypiperidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, 2r,4s-n-boc-4-hydroxypiperidine-2-carboxylic aci, 2r,4s-1-tert-butyl2-methyl4-hydroxypiperidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-cis-4-hydroxy-l-pipecolinic acid methyl ester, 1-o-tert-butyl 2-o-methyl 2r,4s-4-hydroxypiperidine-1,2-dicarboxylate, o1-tert-butyl o2-methyl 2r,4s-4-oxidanylpiperidine-1,2-dicarboxylate, tert-butyl 4s,2r-4-hydroxy-2-methoxycarbonyl piperidinecarboxylate, 2r-4beta-hydroxypiperidine-1,2beta-dicarboxylic acid 1-tert-butyl 2-methyl ester
COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C
259.30
259.3
(2R, 4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, (98% e.e.)
94.0
97.5% min. (GC)
94% min. (HPLC)
C12H21NO5
1g
RNMVWSAJMIKMDY-DTWKUNHWSA-N
1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
2734504
95%,98% ee
Videos
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only