missing translation for 'onlineSavingsMsg'
Learn More

(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide, 97+%

CAS: 638132-66-8 | C18H28OP2 | 322.369 g/mol

$284.22 - $803.22

Chemical Identifiers

CAS 638132-66-8
Molecular Formula C18H28OP2
Molecular Weight (g/mol) 322.369
MDL Number MFCD09750450
InChI Key ORZVYNTUPREFTI-KLHDSHLOSA-N
Synonym r,r-me-bozphos, 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-1-oxide benzene, 1,2-bis 2r,5r-2,5-dimethylphospholano, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, kanata purity, 1,1'-1,2-phenylene bis 2r-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 97+%, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethyl-1??-phospholan-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 1-2-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-yl phenyl-2alpha,5beta-dimethyltetrahydro-1h-phosphole 1-oxide
PubChem CID 11209472
IUPAC Name (2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide
SMILES CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60834MD
View Documents
Thermo Scientific Chemicals
H60834MD
250 mg
Each for $284.22
Only null left
 
AAH6083403
View Documents
Thermo Scientific Chemicals
H6083403
1 g
Each for $803.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

638132-66-8
322.369
ORZVYNTUPREFTI-KLHDSHLOSA-N
11209472
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C
C18H28OP2
MFCD09750450
r,r-me-bozphos, 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-1-oxide benzene, 1,2-bis 2r,5r-2,5-dimethylphospholano, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, kanata purity, 1,1'-1,2-phenylene bis 2r-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 97+%, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethyl-1??-phospholan-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 1-2-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-yl phenyl-2alpha,5beta-dimethyltetrahydro-1h-phosphole 1-oxide
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide

Specifications

638132-66-8
C18H28OP2
250 mg
r,r-me-bozphos, 1-2r,5r-2,5-dimethylphospholanyl-2-2r,5r-2,5-dimethylphospholanyl-1-oxide benzene, 1,2-bis 2r,5r-2,5-dimethylphospholano, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, 1,2-bis 2r,5r-2,5-dimethylphospholano benzene monooxide, kanata purity, 1,1'-1,2-phenylene bis 2r-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 97+%, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethyl-1??-phospholan-1-one, 2r,5r-1-2-2r,5r-2,5-dimethylphospholan-1-yl phenyl-2,5-dimethylphospholane 1-oxide, 1-2-2alpha,5beta-dimethyltetrahydro-1h-phosphole-1-yl phenyl-2alpha,5beta-dimethyltetrahydro-1h-phosphole 1-oxide
−171° (c=1 in Chloroform)
(2R,5R)-1-[2-[(2R,5R)-2,5-dimethylphospholan-1-yl]phenyl]-2,5-dimethyl-1$l^{5}-phospholane 1-oxide
11209472
≥97%
116°C to 122°C
MFCD09750450
Air Sensitive
ORZVYNTUPREFTI-KLHDSHLOSA-N
CC1CCC(P1C2=CC=CC=C2P3(=O)C(CCC3C)C)C
322.369
322.36
(2R,5R)-1-(2-[(2R,5R)-2,5-Dimethylphospholan-1-yl]phenyl)-2,5-dimethylphospholane 1-oxide
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only