missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H52775MD
Description
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid, is used as a novel protected amino acid for the preparation of bestatin and analogs.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
(2S,3R)-3-(Boc-amino)-2-hydroxy-4-phenylbutyric acid | |
C15H21NO5 | |
5909790 | |
Insoluble in water. Soluble in organic solvents. | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(O)C(O)=O | |
250 mg | |
10756280 | |
97% |
62023-65-8 | |
MFCD00057841 | |
2s,3r-3-boc-amino-2-hydroxy-4-phenylbutyric acid, 2s,3r-3-tert-butoxycarbonyl amino-2-hydroxy-4-phenylbutanoic acid, n-boc-2s,3r-2-hydroxy-3-amino-4-phenylbutanoic acid, boc-2s,3r-ahpa, boc-2s,3r-3-amino-2-hydroxy-4-phenylbutyric acid, n-boc-2s,3r-2-hydroxy-3-amino-4-phenylbutanoic a, 2s,3r-2-hydroxy-3-tert-butoxycarbonylamino-4-phenylbutanoic acid, 2s,3r-3-boc-amino-2-hydroxy-4-phenylbutyric acid tlc | |
BHTRKISIDQZUQX-UHFFFAOYNA-N | |
(2S,3R)-2-hydroxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid | |
295.34 | |
295.34 |
Chemical Identifiers
62023-65-8 | |
295.34 | |
BHTRKISIDQZUQX-UHFFFAOYNA-N | |
10756280 |
C15H21NO5 | |
MFCD00057841 | |
2s,3r-3-boc-amino-2-hydroxy-4-phenylbutyric acid, 2s,3r-3-tert-butoxycarbonyl amino-2-hydroxy-4-phenylbutanoic acid, n-boc-2s,3r-2-hydroxy-3-amino-4-phenylbutanoic acid, boc-2s,3r-ahpa, boc-2s,3r-3-amino-2-hydroxy-4-phenylbutyric acid, n-boc-2s,3r-2-hydroxy-3-amino-4-phenylbutanoic a, 2s,3r-2-hydroxy-3-tert-butoxycarbonylamino-4-phenylbutanoic acid, 2s,3r-3-boc-amino-2-hydroxy-4-phenylbutyric acid tlc | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(O)C(O)=O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only