missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2S,3S)-3-Methylpyrrolidine-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 10512-89-7
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
InChI Key CNPSFBUUYIVHAP-WHFBIAKZSA-N
Synonym 2s,3s-3-methylpyrrolidine-2-carboxylic acid, trans-3-methyl-l-proline, l-proline, 3-methyl-, 3s, trans-3-methylproline, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid
PubChem CID 2724879
IUPAC Name (2S,3S)-3-methylpyrrolidine-2-carboxylic acid
SMILES CC1CCNC1C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC290092500
SDS View Documents
Thermo Scientific Chemicals
290092500
250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC290090010
SDS View Documents
Thermo Scientific Chemicals
290090010
1g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

10512-89-7
129.16
2s,3s-3-methylpyrrolidine-2-carboxylic acid, trans-3-methyl-l-proline, l-proline, 3-methyl-, 3s, trans-3-methylproline, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid
(2S,3S)-3-methylpyrrolidine-2-carboxylic acid
C6H11NO2
CNPSFBUUYIVHAP-WHFBIAKZSA-N
2724879
CC1CCNC1C(=O)O

Specifications

10512-89-7
97%
C6H11NO2
− 28.00 (25.00°C c=0.7,water)
2s,3s-3-methylpyrrolidine-2-carboxylic acid, trans-3-methyl-l-proline, l-proline, 3-methyl-, 3s, trans-3-methylproline, 3beta-methyl-l-proline, 3s-3-methyl-l-proline, h-l-trans-pro 3-me-oh, 2s,3s-3-methylpyrrolidine-2-carboxylicacid, l-trans-3-methylproline, 2s,3s-3-methyl-pyrroline-2-carboxylic acid
CC1CCNC1C(=O)O
129.16
129.16
(2S, 3S)-3-Methylpyrrolidine-2-carboxylic acid
Authentic
Glass bottle
250mg
− 28.00
CNPSFBUUYIVHAP-WHFBIAKZSA-N
(2S,3S)-3-methylpyrrolidine-2-carboxylic acid
2724879
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only