missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(2-Furyl)acrolein 98.0+%, TCI America™

Supplier:  TCI America F018125G

Catalog No. F018125G

Explore available promotions

Description

Description

Specifications

Specifications

3-(2-Furyl)acrolein
54°C
C7H6O2
25 g
3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein
C1=COC(=C1)C=CC=O
122.123
122.12
Crystalline Powder
623-30-3
143°C
MFCD00003256
1759
VZIRCHXYMBFNFD-HNQUOIGGSA-N
(E)-3-(furan-2-yl)prop-2-enal
1549521
≥98.0% (GC)

Chemical Identifiers

623-30-3
122.123
VZIRCHXYMBFNFD-HNQUOIGGSA-N
1549521
C1=COC(=C1)C=CC=O
C7H6O2
MFCD00003256
3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein
(E)-3-(furan-2-yl)prop-2-enal
Videos
Safety and Handling

Safety and Handling

RTECSNumber : LT8528500

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions