Learn More
3-(2-Methoxyphenoxy)-1,2-propanediol, 98+%, Thermo Scientific Chemicals
$67.74 - $195.40
Chemical Identifiers
CAS | 93-14-1 |
---|---|
Molecular Formula | C10H14O4 |
Molecular Weight (g/mol) | 198.22 |
MDL Number | MFCD00016873 |
InChI Key | HSRJKNPTNIJEKV-UHFFFAOYNA-N |
Synonym | guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol |
PubChem CID | 3516 |
IUPAC Name | 3-(2-methoxyphenoxy)propane-1,2-diol |
SMILES | COC1=CC=CC=C1OCC(O)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1682718
|
Thermo Scientific Chemicals
A1682718 |
50 g |
Each for $67.74
|
|
|||||
AAA1682730
|
Thermo Scientific Chemicals
A1682730 |
250 g |
Each for $195.40
|
|
|||||
Description
3-(2-Methoxyphenoxy)-1,2-propanediol is involved in centrally acting muscle relaxant with expectorant properties. It is often used as anesthetic during animal surgery. It is used in the treatment of coughing
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (50 mg/ml at 25°C), methanol, DMSO (40 mg/ml at 25°C), DMF, ethanol (50 mg/ml), glycerol and glycol.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Chemical Identifiers
93-14-1 | |
198.22 | |
HSRJKNPTNIJEKV-UHFFFAOYNA-N | |
3516 | |
COC1=CC=CC=C1OCC(O)CO |
C10H14O4 | |
MFCD00016873 | |
guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol | |
3-(2-methoxyphenoxy)propane-1,2-diol |
Specifications
93-14-1 | |
215°C (19 mmHg) | |
MFCD00016873 | |
2049375 | |
guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol | |
HSRJKNPTNIJEKV-UHFFFAOYNA-N | |
3-(2-methoxyphenoxy)propane-1,2-diol | |
3516 | |
≥99% |
80°C to 82°C | |
C10H14O4 | |
50 g | |
14,4555 | |
Soluble in water (50mg/ml at 25°C),methanol,DMSO (40mg/ml at 25°C),DMF,ethanol (50mg/ml),glycerol and glycol. | |
COC1=CC=CC=C1OCC(O)CO | |
198.22 | |
198.22 | |
3-(2-Methoxyphenoxy)-1,2-propanediol |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 202-222-5
RTECSNumber : TY8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only