missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
-
Home
-
Products
-
3-(2-Naphthyl)-L-alanine, 98%, Thermo Scientific™
3-(2-Naphthyl)-L-alanine, 98%, Thermo Scientific™
Chemical Identifiers
CAS |
58438-03-2 |
Molecular Formula |
C13H13NO2 |
Molecular Weight (g/mol) |
215.25 |
MDL Number |
MFCD00066087 |
InChI Key |
JPZXHKDZASGCLU-LBPRGKRZSA-N |
Synonym |
3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid |
PubChem CID |
185915 |
IUPAC Name |
(2S)-2-amino-3-naphthalen-2-ylpropanoic acid |
SMILES |
N[C@@H](CC1=CC=C2C=CC=CC2=C1)C(O)=O |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
58438-03-2 |
|
215.25 |
|
JPZXHKDZASGCLU-LBPRGKRZSA-N |
|
185915 |
|
N[C@@H](CC1=CC=C2C=CC=CC2=C1)C(O)=O |
|
C13H13NO2 |
|
MFCD00066087 |
|
3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid |
|
(2S)-2-amino-3-naphthalen-2-ylpropanoic acid |
Specifications
|
58438-03-2 |
|
100.0 |
|
C13H13NO2 |
|
MFCD00066087 |
|
3-2-naphthyl-l-alanine, h-2-nal-oh, s-2-amino-3-naphthalen-2-yl propanoic acid, l-2-naphthylalanine, beta-2-naphthyl-alanine, unii-w425q6kv9r, l-3-2-naphthyl-alanine, nal, 2s-2-amino-3-2-naphthyl propanoic acid, s-2-amino-3-naphthalen-2-yl-propionic acid |
|
N[C@@H](CC1=CC=C2C=CC=CC2=C1)C(O)=O |
|
215.25 |
|
215.25 |
|
250mg |
|
97.5 |
|
98% |
|
Authentic |
|
Glass bottle |
|
JPZXHKDZASGCLU-LBPRGKRZSA-N |
|
(2S)-2-amino-3-naphthalen-2-ylpropanoic acid |
|
185915 |
|
≥97.5% |
|
3-(2-Naphthyl)-L-alanine |
SDS
missing translation for 'documents'
RUO â Research Use Only