missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3',4',7-Tetrahydroxyflavone, 96%, Thermo Scientific Chemicals

$132.58 - $354.27

Chemical Identifiers

CAS 528-48-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
MDL Number MFCD00006829
InChI Key XHEFDIBZLJXQHF-UHFFFAOYSA-N
Synonym fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
PubChem CID 5281614
ChEBI CHEBI:42567
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL14207MD
View Documents
Thermo Scientific Chemicals
L14207MD
250 mg
Each for $132.58
 
AAL1420703
View Documents
Thermo Scientific Chemicals
L1420703
1 g
Each for $354.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

528-48-3
286.239
XHEFDIBZLJXQHF-UHFFFAOYSA-N
5281614
2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
C15H10O6
MFCD00006829
fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
CHEBI:42567
C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O

Specifications

528-48-3
C15H10O6
250 mg
14,4088
XHEFDIBZLJXQHF-UHFFFAOYSA-N
2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
5281614
286.24
3,3',4',7-Tetrahydroxyflavone
330°C (decomposition)
MFCD00006829
292829
fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O
286.239
CHEBI:42567
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-434-4

RTECSNumber : LK9250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only