missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)propionic acid, 97%

CAS: 42287-90-1 | C9H9BrO2 | 229.073 g/mol

$54.56 - $173.36

Chemical Identifiers

CAS 42287-90-1
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD01310792
InChI Key DWKWMFSWLCIMKI-UHFFFAOYSA-N
Synonym 3-3-bromophenyl propanoic acid, 3-3-bromophenyl propionic acid, 3-3-bromo-phenyl-propionic acid, 3-bromophenylpropanoic acid, acmc-209jor, m-bromohydrocinnamic acid, 3-bromohydrocinnamic acid, benzenepropanoic acid,3-bromo, 3-3-bromophenyl propionicacid, 3-m-bromophenyl propionic acid
PubChem CID 2063862
IUPAC Name 3-(3-bromophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)Br)CCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1808003
View Documents
Thermo Scientific Chemicals
L1808003
1 g
Each for $54.56
Only null left
 
AAL1808006
View Documents
Thermo Scientific Chemicals
L1808006
5 g
Each for $173.36
Only null left
 
Description

Description

It is used in the synthesis and characterization of monoisomeric 1,8,15,22-substituted (A3B and A2B2) phthalocyanines and phthalocyanine-fullerene dyads.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis and characterization of monoisomeric 1,8,15,22-substituted (A3B and A2B2) phthalocyanines and phthalocyanine-fullerene dyads.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

42287-90-1
229.073
DWKWMFSWLCIMKI-UHFFFAOYSA-N
2063862
C1=CC(=CC(=C1)Br)CCC(=O)O
C9H9BrO2
MFCD01310792
3-3-bromophenyl propanoic acid, 3-3-bromophenyl propionic acid, 3-3-bromo-phenyl-propionic acid, 3-bromophenylpropanoic acid, acmc-209jor, m-bromohydrocinnamic acid, 3-bromohydrocinnamic acid, benzenepropanoic acid,3-bromo, 3-3-bromophenyl propionicacid, 3-m-bromophenyl propionic acid
3-(3-bromophenyl)propanoic acid

Specifications

42287-90-1
C9H9BrO2
1 g
3-3-bromophenyl propanoic acid, 3-3-bromophenyl propionic acid, 3-3-bromo-phenyl-propionic acid, 3-bromophenylpropanoic acid, acmc-209jor, m-bromohydrocinnamic acid, 3-bromohydrocinnamic acid, benzenepropanoic acid,3-bromo, 3-3-bromophenyl propionicacid, 3-m-bromophenyl propionic acid
DWKWMFSWLCIMKI-UHFFFAOYSA-N
3-(3-bromophenyl)propanoic acid
2063862
97%
63°C to 65°C
MFCD01310792
2362753
Insoluble in water.
C1=CC(=CC(=C1)Br)CCC(=O)O
229.073
229.07
3-(3-Bromophenyl)propionic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only