missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3-Dimethyl-1-butene 96.0+%, TCI America™
$111.84 - $389.59
Chemical Identifiers
| CAS | 558-37-2 |
|---|---|
| Molecular Formula | C6H12 |
| Molecular Weight (g/mol) | 84.16 |
| MDL Number | MFCD00008853 |
| InChI Key | PKXHXOTZMFCXSH-UHFFFAOYSA-N |
| Synonym | 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk |
| PubChem CID | 11210 |
| IUPAC Name | 3,3-dimethylbut-1-ene |
| SMILES | CC(C)(C)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D116325ML
|
TCI America
D116325ML |
25 mL |
Each for $111.84
|
|
|||||
|
D1163100ML
|
TCI America
D1163100ML |
100 mL |
Each for $389.59
|
|
|||||
Chemical Identifiers
| 558-37-2 | |
| 84.16 | |
| PKXHXOTZMFCXSH-UHFFFAOYSA-N | |
| 11210 | |
| CC(C)(C)C=C |
| C6H12 | |
| MFCD00008853 | |
| 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk | |
| 3,3-dimethylbut-1-ene |
Specifications
| 558-37-2 | |
| 41°C | |
| MFCD00008853 | |
| 3295 | |
| PKXHXOTZMFCXSH-UHFFFAOYSA-N | |
| 3,3-dimethylbut-1-ene | |
| 11210 | |
| ≥96.0% (GC) | |
| 3,3-Dimethyl-1-butene |
| Colorless | |
| C6H12 | |
| 25 mL | |
| 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk | |
| CC(C)(C)C=C | |
| 84.16 | |
| 84.16 | |
| Liquid |
Safety and Handling
TSCA : Yes