missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butyne, 98%

CAS: 917-92-0 | C6H10 | 82.146 g/mol

$167.18 - $492.52

Chemical Identifiers

CAS 917-92-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008852
InChI Key PPWNCLVNXGCGAF-UHFFFAOYSA-N
PubChem CID 13512
IUPAC Name 3,3-dimethylbut-1-yne
SMILES CC(C)(C)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1401306
View Documents
Thermo Scientific Chemicals
A1401306
5 g
Each for $167.18
Only null left
 
AAA1401314
View Documents
Thermo Scientific Chemicals
A1401314
25 g
Each for $492.52
Only null left
 
Description

Description

3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butyne is used as an active pharmaceutical intermediate for terbinafine. It is also used in the preparation of 2,2,7,7-tetramethyl-octa-3,5-diyne. Further, it is used in the synthesis of erythro and threo isomers of B-(3,3-dimethyl-1,2-dideuterio-1-butyl)-9-BBN by hydroboration-deuteronolysis-hydroboration sequence.

Solubility
Miscible with chloroform, benzene and toluene. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

917-92-0
82.146
PPWNCLVNXGCGAF-UHFFFAOYSA-N
3,3-dimethylbut-1-yne
C6H10
MFCD00008852
13512
CC(C)(C)C#C

Specifications

917-92-0
0.671
<−34°C (−29°F)
C6H10
(CH3)3CC≡CH
5 g
1697100
Miscible with chloroform,benzene and toluene. Immiscible with water.
CC(C)(C)C#C
82.146
82.15
3,3-Dimethyl-1-butyne
-78°C
37°C to 38°C
Recognizable
1.3745
MFCD00008852
UN3295
3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
PPWNCLVNXGCGAF-UHFFFAOYSA-N
3,3-dimethylbut-1-yne
13512
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: I

EINECSNumber : 213-035-3

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only