missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 131251000

 View more versions of this product

Catalog No. AC131251000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3, 3-Dimethyl-2-butanone
75-97-8
Colorless to Yellow
106.0°C
Authentic
Glass Bottle
1.3950 to 1.3980
MFCD00008846
01, 694
15, 7551
Solubility in water: very soluble. Other solubilities: soluble in alcohol,ether and acetone
CC(=O)C(C)(C)C
100.16
100.16
Liquid
96%
-52.5°C
0.8000g/mL
5°C
95% min. (GC)
C6H12O
(CH3)3CCOCH3
100 mL
0.8
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
PJGSXYOJTGTZAV-UHFFFAOYSA-N
3,3-dimethylbutan-2-one
6416
96%

Chemical Identifiers

75-97-8
100.16
PJGSXYOJTGTZAV-UHFFFAOYSA-N
6416
CC(=O)C(C)(C)C
C6H12O
MFCD00008846
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
3,3-dimethylbutan-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-920-4

RTECSNumber : EL7700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only