Learn More
3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131255000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3, 3-Dimethyl-2-butanone | |
75-97-8 | |
Colorless to Yellow | |
106.0°C | |
Authentic | |
Glass bottle | |
1.3950 to 1.3980 | |
MFCD00008846 | |
01, 694 | |
15, 7551 | |
Solubility in water: very soluble. Other solubilities: soluble in alcohol,ether and acetone | |
CC(=O)C(C)(C)C | |
100.16 | |
100.16 | |
Liquid |
96% | |
-52.5°C | |
0.8000g/mL | |
5°C | |
95% min. (GC) | |
C6H12O | |
(CH3)3CCOCH3 | |
500 mL | |
0.8 | |
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone | |
PJGSXYOJTGTZAV-UHFFFAOYSA-N | |
3,3-dimethylbutan-2-one | |
6416 | |
96% |
Chemical Identifiers
75-97-8 | |
100.16 | |
PJGSXYOJTGTZAV-UHFFFAOYSA-N | |
6416 | |
CC(=O)C(C)(C)C |
C6H12O | |
MFCD00008846 | |
pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone | |
3,3-dimethylbutan-2-one |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 200-920-4
RTECSNumber : EL7700000
TSCA : TSCA
RUO – Research Use Only