missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethylazobenzene 98.0+%, TCI America™

$150.98 - $456.70

Chemical Identifiers

CAS 588-04-5
Molecular Formula C14H14N2
Molecular Weight (g/mol) 210.28
MDL Number MFCD00048080
InChI Key HPSZIXYLILUGIP-UHFFFAOYSA-N
Synonym m-Azotoluene
PubChem CID 11491
IUPAC Name bis(3-methylphenyl)diazene
SMILES CC1=CC(=CC=C1)N=NC2=CC=CC(=C2)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D06761G
View Documents
TCI America
D06761G
1 g N/A
Only null left
 
D06765G
View Documents
TCI America
D06765G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

588-04-5
210.28
HPSZIXYLILUGIP-UHFFFAOYSA-N
11491
CC1=CC(=CC=C1)N=NC2=CC=CC(=C2)C
C14H14N2
MFCD00048080
m-Azotoluene
bis(3-methylphenyl)diazene

Specifications

588-04-5
C14H14N2
1 g
HPSZIXYLILUGIP-UHFFFAOYSA-N
bis(3-methylphenyl)diazene
11491
≥98.0% (GC)
3,3′-Dimethylazobenzene
53°C
MFCD00048080
m-Azotoluene
CC1=CC(=CC=C1)N=NC2=CC=CC(=C2)C
210.28
210.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'