missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,3-Dimethylglutarimide 95.0+%, TCI America™
Supplier: TCI America D132410G
Specifications
3,3-Dimethylglutarimide | |
146°C | |
C7H11NO2 | |
10 g | |
YUJCWMGBRDBPDL-UHFFFAOYSA-N | |
4,4-dimethylpiperidine-2,6-dione | |
14292 | |
≥95.0% (N) |
1123-40-6 | |
White-Yellow | |
MFCD00006671 | |
3,3-dimethylglutarimide, 2,6-piperidinedione, 4,4-dimethyl, glutarimide, 3,3-dimethyl, 4,4-dimethyl-2,6-piperidinedione, beta,beta-dimethylglutarimide, 3,3-dimethyl-glutarimide, .beta.,.beta.-dimethylglutarimide, 4,6-piperidinedione, 2, 4,4-dimethyl, glutarimide,3-dimethyl | |
CC1(CC(=O)NC(=O)C1)C | |
141.17 | |
141.17 | |
Crystalline Powder |
Chemical Identifiers
1123-40-6 | |
141.17 | |
YUJCWMGBRDBPDL-UHFFFAOYSA-N | |
14292 | |
CC1(CC(=O)NC(=O)C1)C |
C7H11NO2 | |
MFCD00006671 | |
3,3-dimethylglutarimide, 2,6-piperidinedione, 4,4-dimethyl, glutarimide, 3,3-dimethyl, 4,4-dimethyl-2,6-piperidinedione, beta,beta-dimethylglutarimide, 3,3-dimethyl-glutarimide, .beta.,.beta.-dimethylglutarimide, 4,6-piperidinedione, 2, 4,4-dimethyl, glutarimide,3-dimethyl | |
4,4-dimethylpiperidine-2,6-dione |
Safety and Handling
RTECSNumber : MA4348000
TSCA : No