missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3-Dimethylheptane 98.0+%, TCI America™

Supplier:  TCI America D12061ML

Catalog No. D12061ML

Explore available promotions

Description

Description

Specifications

Specifications

3,3-Dimethylheptane
Colorless
C9H20
1 mL
heptane, 3,3-dimethyl, 3,3-dimethylheptane, acmc-1aeuh, 3,3-dimethyl heptane
CCCCC(C)(C)CC
128.259
128.26
Liquid
4032-86-4
137°C
MFCD00048811
1920
BVAKDOXCVSMKHE-UHFFFAOYSA-N
3,3-dimethylheptane
520991
≥98.0% (GC)

Chemical Identifiers

4032-86-4
128.259
BVAKDOXCVSMKHE-UHFFFAOYSA-N
520991
CCCCC(C)(C)CC
C9H20
MFCD00048811
heptane, 3,3-dimethyl, 3,3-dimethylheptane, acmc-1aeuh, 3,3-dimethyl heptane
3,3-dimethylheptane
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0009

TSCA : No

SDS
Documents

Documents

Promotions

Promotions