Learn More
3-(3-Furyl)acrylic acid, 98%
CAS: 39244-10-5 | C7H6O3 | 138.12 g/mol
$114.74 - $1774.14
Chemical Identifiers
| CAS | 39244-10-5 |
|---|---|
| Molecular Formula | C7H6O3 |
| Molecular Weight (g/mol) | 138.12 |
| MDL Number | MFCD00075074 |
| InChI Key | JHAPZUDWRRBZHZ-UHFFFAOYSA-N |
| Synonym | 3-3-furyl acrylic acid, 3-furan-3-yl acrylic acid, trans-3-furanacrylic acid, e-3-furan-3-yl acrylic acid, 2-propenoic acid, 3-3-furanyl-, 2e, 2e-3-3-furyl prop-2-enoic acid, 2e-3-furan-3-yl prop-2-enoic acid, 3-3-furanyl-2-propenoic acid, 2e-3-3-furyl acrylic acid, furan-3-acrylic acid |
| PubChem CID | 643403 |
| IUPAC Name | (E)-3-(furan-3-yl)prop-2-enoic acid |
| SMILES | OC(=O)C=CC1=COC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1314303
|
Thermo Scientific Chemicals
L1314303 |
1 g |
Each for $114.74
|
|
|||||
|
AAL1314306
|
Thermo Scientific Chemicals
L1314306 |
5 g |
Each for $448.38
|
|
|||||
|
AAL1314314
|
Thermo Scientific Chemicals
L1314314 |
25 g |
Each for $1,774.14
|
|
|||||
Description
3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(3-Furyl)acrylic acid is used as an intermediate in chemical research and pharmaceuticals.
Solubility
Sparingly soluble in water (0.56 g/L at 25°C).
Notes
Store at room temperature. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 39244-10-5 | |
| 138.12 | |
| JHAPZUDWRRBZHZ-UHFFFAOYSA-N | |
| 643403 | |
| OC(=O)C=CC1=COC=C1 |
| C7H6O3 | |
| MFCD00075074 | |
| 3-3-furyl acrylic acid, 3-furan-3-yl acrylic acid, trans-3-furanacrylic acid, e-3-furan-3-yl acrylic acid, 2-propenoic acid, 3-3-furanyl-, 2e, 2e-3-3-furyl prop-2-enoic acid, 2e-3-furan-3-yl prop-2-enoic acid, 3-3-furanyl-2-propenoic acid, 2e-3-3-furyl acrylic acid, furan-3-acrylic acid | |
| (E)-3-(furan-3-yl)prop-2-enoic acid |
Specifications
| 39244-10-5 | |
| C7H6O3 | |
| 1 g | |
| 3-3-furyl acrylic acid, 3-furan-3-yl acrylic acid, trans-3-furanacrylic acid, e-3-furan-3-yl acrylic acid, 2-propenoic acid, 3-3-furanyl-, 2e, 2e-3-3-furyl prop-2-enoic acid, 2e-3-furan-3-yl prop-2-enoic acid, 3-3-furanyl-2-propenoic acid, 2e-3-3-furyl acrylic acid, furan-3-acrylic acid | |
| JHAPZUDWRRBZHZ-UHFFFAOYSA-N | |
| (E)-3-(furan-3-yl)prop-2-enoic acid | |
| 643403 | |
| 98% |
| 152°C to 154°C | |
| MFCD00075074 | |
| 1306450 | |
| Sparingly soluble in water (0.56g/L at 25°C). | |
| OC(=O)C=CC1=COC=C1 | |
| 138.12 | |
| 138.12 | |
| 3-(3-Furyl)acrylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only