Learn More
3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals
$188.10 - $682.99
Chemical Identifiers
CAS | 621-54-5 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.176 |
MDL Number | MFCD00016554 |
InChI Key | QVWAEZJXDYOKEH-UHFFFAOYSA-N |
Synonym | 3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl propionic acid, dihydro-3-coumaric acid, benzenepropanoic acid, 3-hydroxy, 3-m-hydroxyphenyl propionic acid, 3-hydroxybenzenepropanoic acid, 3-hydroxyphenylpropionic acid, m-hydroxyphenylpropionic acid, beta-m-hydroxyphenyl propionic acid, hydrocinnamic acid, m-hydroxy |
PubChem CID | 91 |
ChEBI | CHEBI:1427 |
IUPAC Name | 3-(3-hydroxyphenyl)propanoic acid |
SMILES | C1=CC(=CC(=C1)O)CCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0127906
|
Thermo Scientific Chemicals
L0127906 |
5 g |
Each for $188.10
|
|
||||||
AAL0127914
|
Thermo Scientific Chemicals
L0127914 |
25 g |
Each for $682.99
|
|
||||||
Description
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.
Solubility
Soluble in chloroform and methanol.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
Chemical Identifiers
621-54-5 | |
166.176 | |
QVWAEZJXDYOKEH-UHFFFAOYSA-N | |
91 | |
3-(3-hydroxyphenyl)propanoic acid |
C9H10O3 | |
MFCD00016554 | |
3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl propionic acid, dihydro-3-coumaric acid, benzenepropanoic acid, 3-hydroxy, 3-m-hydroxyphenyl propionic acid, 3-hydroxybenzenepropanoic acid, 3-hydroxyphenylpropionic acid, m-hydroxyphenylpropionic acid, beta-m-hydroxyphenyl propionic acid, hydrocinnamic acid, m-hydroxy | |
CHEBI:1427 | |
C1=CC(=CC(=C1)O)CCC(=O)O |
Specifications
621-54-5 | |
C9H10O3 | |
5 g | |
3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl propionic acid, dihydro-3-coumaric acid, benzenepropanoic acid, 3-hydroxy, 3-m-hydroxyphenyl propionic acid, 3-hydroxybenzenepropanoic acid, 3-hydroxyphenylpropionic acid, m-hydroxyphenylpropionic acid, beta-m-hydroxyphenyl propionic acid, hydrocinnamic acid, m-hydroxy | |
QVWAEZJXDYOKEH-UHFFFAOYSA-N | |
3-(3-hydroxyphenyl)propanoic acid | |
91 | |
166.18 | |
3-(3-Hydroxyphenyl)propionic acid |
110°C to 114°C | |
MFCD00016554 | |
1947445 | |
Soluble in chloroform and methanol. | |
C1=CC(=CC(=C1)O)CCC(=O)O | |
166.176 | |
CHEBI:1427 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-692-8
RTECSNumber : MW5340000
TSCA : No
RUO – Research Use Only