missing translation for 'onlineSavingsMsg'
Learn More

[3-(3-Methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 916766-84-2
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD09817471
InChI Key ZBIYTANHMLGZFB-UHFFFAOYSA-N
Synonym 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanol, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl alcohol, 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methan-1-ol, benzenemethanol,3-3-methyl-1,2,4-oxadiazol-5-yl
PubChem CID 24229498
IUPAC Name [3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol
SMILES CC1=NOC(=N1)C2=CC(=CC=C2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC49509CB
SDS View Documents
Thermo Scientific Chemicals
CC49509CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC49509DA
SDS View Documents
Thermo Scientific Chemicals
CC49509DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

916766-84-2
190.202
ZBIYTANHMLGZFB-UHFFFAOYSA-N
24229498
CC1=NOC(=N1)C2=CC(=CC=C2)CO
C10H10N2O2
MFCD09817471
3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanol, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl alcohol, 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methan-1-ol, benzenemethanol,3-3-methyl-1,2,4-oxadiazol-5-yl
[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol

Specifications

916766-84-2
100.0
Amber Glass Bottle
MFCD09817471
3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanol, 3-3-methyl-1,2,4-oxadiazol-5-yl benzyl alcohol, 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methan-1-ol, benzenemethanol,3-3-methyl-1,2,4-oxadiazol-5-yl
CC1=NOC(=N1)C2=CC(=CC=C2)CO
190.202
190.2
[3-(3-Methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methanol
96.5
97%
C10H10N2O2
250mg
ZBIYTANHMLGZFB-UHFFFAOYSA-N
[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol
24229498
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'