missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(3-Pyridyl)propionic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1400706

 View more versions of this product

Catalog No. AAL1400706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(3-Pyridyl)propionic acid
156°C to 159°C
MFCD00274197
122232
WDGXIUUWINKTGP-UHFFFAOYSA-N
3-pyridin-3-ylpropanoic acid
259624
97%
3724-19-4
C8H9NO2
5 g
3-3-pyridyl propionic acid, 3-pyridinepropionic acid, 3-pyridinepropanoic acid, 3-pyridin-3-yl propanoic acid, 3-3-pyridyl propionicacid, pyridine-3-propionic acid, 3-3-pyridyl propanoic acid, 3-pyridin-3-yl propionic acid, pubchem8701, maybridge4_001332
C1=CC(=CN=C1)CCC(=O)O
151.165
151.17

Chemical Identifiers

3724-19-4
151.165
WDGXIUUWINKTGP-UHFFFAOYSA-N
259624
C1=CC(=CN=C1)CCC(=O)O
C8H9NO2
MFCD00274197
3-3-pyridyl propionic acid, 3-pyridinepropionic acid, 3-pyridinepropanoic acid, 3-pyridin-3-yl propanoic acid, 3-3-pyridyl propionicacid, pyridine-3-propionic acid, 3-3-pyridyl propanoic acid, 3-pyridin-3-yl propionic acid, pubchem8701, maybridge4_001332
3-pyridin-3-ylpropanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only