Learn More
3-(3-Pyridyl)propionic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1400714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-(3-Pyridyl)propionic acid | |
156°C to 159°C | |
MFCD00274197 | |
122232 | |
WDGXIUUWINKTGP-UHFFFAOYSA-N | |
3-pyridin-3-ylpropanoic acid | |
259624 | |
97% |
3724-19-4 | |
C8H9NO2 | |
25 g | |
3-3-pyridyl propionic acid, 3-pyridinepropionic acid, 3-pyridinepropanoic acid, 3-pyridin-3-yl propanoic acid, 3-3-pyridyl propionicacid, pyridine-3-propionic acid, 3-3-pyridyl propanoic acid, 3-pyridin-3-yl propionic acid, pubchem8701, maybridge4_001332 | |
C1=CC(=CN=C1)CCC(=O)O | |
151.165 | |
151.17 |
Chemical Identifiers
3724-19-4 | |
151.165 | |
WDGXIUUWINKTGP-UHFFFAOYSA-N | |
259624 | |
C1=CC(=CN=C1)CCC(=O)O |
C8H9NO2 | |
MFCD00274197 | |
3-3-pyridyl propionic acid, 3-pyridinepropionic acid, 3-pyridinepropanoic acid, 3-pyridin-3-yl propanoic acid, 3-3-pyridyl propionicacid, pyridine-3-propionic acid, 3-3-pyridyl propanoic acid, 3-pyridin-3-yl propionic acid, pubchem8701, maybridge4_001332 | |
3-pyridin-3-ylpropanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only