missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3',4',5,7-Tetrahydroxyflavone 98.0+%, TCI America™
Supplier: TCI America T26825G
Specifications
3′,4′,5,7-Tetrahydroxyflavone | |
330°C | |
MFCD00017309 | |
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide | |
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O | |
286.239 | |
CHEBI:15864 | |
≥98.0% (HPLC) |
491-70-3 | |
C15H10O6 | |
5 g | |
IQPNAANSBPBGFQ-UHFFFAOYSA-N | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one | |
5280445 | |
286.24 | |
Crystalline Powder |
Chemical Identifiers
491-70-3 | |
286.239 | |
IQPNAANSBPBGFQ-UHFFFAOYSA-N | |
5280445 | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one |
C15H10O6 | |
MFCD00017309 | |
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide | |
CHEBI:15864 | |
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O |
Safety and Handling
RTECSNumber : LK9275210
TSCA : No
Recommended Storage : Refrigerator