missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4,5-Trifluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™
Supplier: TCI America T32605G
Specifications
3,4,5-Trifluoro-4′-(trans-4-propylcyclohexyl)biphenyl | |
42°C | |
C21H23F3 | |
5 g | |
RRKRBRVTKIRLHH-UHFFFAOYSA-N | |
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene | |
15014921 | |
≥98.0% (GC) |
132123-39-8 | |
White | |
MFCD09839001 | |
3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl | |
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F | |
332.41 | |
332.41 | |
Crystalline Powder |
Chemical Identifiers
132123-39-8 | |
332.41 | |
RRKRBRVTKIRLHH-UHFFFAOYSA-N | |
15014921 | |
CCCC1CCC(CC1)C2=CC=C(C=C2)C3=CC(=C(C(=C3)F)F)F |
C21H23F3 | |
MFCD09839001 | |
3,4,5-trifluoro-4'-trans-4-propylcyclohexyl-1,1'-biphenyl, 4'-trans-4-propylcyclohexyl-3,4,5-trifluorobiphenyl, 1,2,3-trifluoro-5-4-4-propylcyclohexyl phenyl benzene, 4'-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, 3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, trans-3,4,5-trifluoro-4'-4-propylcyclohexyl biphenyl, 4-trans-4-propylcyclohexyl-3,4,5-trifluoro-biphenyl, trans-3,4,5-trifluoro-4'-4-n-propylcyclohexyl biphenyl, 1,1'-biphenyl, 3,4,5-trifluoro-4'-trans-4-propylcyclohexyl | |
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)phenyl]benzene |
Safety and Handling
EINECSNumber : (4)-1980
TSCA : No