missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trihydroxybenzaldehyde 98.0+%, TCI America™

Supplier:  TCI America T26501G

 View more versions of this product

Catalog No. T26501G


Explore available promotions

Description

Description

Specifications

Specifications

3,4,5-Trihydroxybenzaldehyde
215°C
MFCD00003371
gallaldehyde, benzaldehyde, 3,4,5-trihydroxy, pyrogallol-5-carboxaldehyde, 3,4,5-trihydroxy benzaldehyde, pubchem21300, 3,4,5-trihydroxybenz, acmc-209c9s, 3,4,5-trihydroxybenzaldhyde, 3,4,5-trihydroxy-benzaldehyde, 3,4,5-tris oxidanyl benzaldehyde
OC1=CC(C=O)=CC(O)=C1O
154.12
154.12
Crystalline Powder
13677-79-7
C7H6O4
1 g
RGZHEOWNTDJLAQ-UHFFFAOYSA-N
3,4,5-trihydroxybenzaldehyde
83651
≥98.0% (GC,T)

Chemical Identifiers

13677-79-7
154.12
RGZHEOWNTDJLAQ-UHFFFAOYSA-N
83651
OC1=CC(C=O)=CC(O)=C1O
C7H6O4
MFCD00003371
gallaldehyde, benzaldehyde, 3,4,5-trihydroxy, pyrogallol-5-carboxaldehyde, 3,4,5-trihydroxy benzaldehyde, pubchem21300, 3,4,5-trihydroxybenz, acmc-209c9s, 3,4,5-trihydroxybenzaldhyde, 3,4,5-trihydroxy-benzaldehyde, 3,4,5-tris oxidanyl benzaldehyde
3,4,5-trihydroxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions