missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzaldehyde, 98%

CAS: 86-81-7 | C10H12O4 | 196.202 g/mol

$62.67 - $568.42

Chemical Identifiers

CAS 86-81-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003364
InChI Key OPHQOIGEOHXOGX-UHFFFAOYSA-N
PubChem CID 6858
IUPAC Name 3,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1OC)OC)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1503814
View Documents
Thermo Scientific Chemicals
A1503814
25 g
Each for $62.67
Only null left
 
AAA1503822
View Documents
Thermo Scientific Chemicals
A1503822
100 g
Each for $171.40
Only null left
 
AAA1503836
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1503836
500 g
Each for $568.42
Only null left
 
Description

Description

3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

86-81-7
196.202
OPHQOIGEOHXOGX-UHFFFAOYSA-N
3,4,5-trimethoxybenzaldehyde
C10H12O4
MFCD00003364
6858
COC1=CC(=CC(=C1OC)OC)C=O

Specifications

86-81-7
163°C to 165°C (10 mmHg)
MFCD00003364
395163
benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde
OPHQOIGEOHXOGX-UHFFFAOYSA-N
3,4,5-trimethoxybenzaldehyde
6858
98%
73°C to 77°C
C10H12O4
25 g
Air Sensitive
Soluble in water (1.49mg/L at 25°C),alcohol,toluene,acetone,and methanol.
COC1=CC(=CC(=C1OC)OC)C=O
196.202
196.2
3,4,5-Trimethoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 201-701-6

RTECSNumber : CU8462000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only