Learn More
3,4,5-Trimethoxybenzylamine, 96%, Thermo Scientific Chemicals
$134.18 - $134.18
Chemical Identifiers
CAS | 18638-99-8 |
---|---|
Molecular Formula | C10H15NO3 |
Molecular Weight (g/mol) | 197.24 |
MDL Number | MFCD00017150 |
InChI Key | YUPUSBMJCFBHAP-UHFFFAOYSA-N |
Synonym | 3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine |
PubChem CID | 87736 |
IUPAC Name | (3,4,5-trimethoxyphenyl)methanamine |
SMILES | COC1=CC(=CC(=C1OC)OC)CN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC351170050
|
Thermo Scientific Chemicals
351170050 |
5 g | Glass bottle |
Each for $134.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18638-99-8 | |
197.24 | |
YUPUSBMJCFBHAP-UHFFFAOYSA-N | |
87736 | |
COC1=CC(=CC(=C1OC)OC)CN |
C10H15NO3 | |
MFCD00017150 | |
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine | |
(3,4,5-trimethoxyphenyl)methanamine |
Specifications
18638-99-8 | |
100.0 | |
1.1550g/mL | |
Authentic | |
Glass bottle | |
1.5455 to 1.5485 | |
MFCD00017150 | |
1.155 | |
YUPUSBMJCFBHAP-UHFFFAOYSA-N | |
(3,4,5-trimethoxyphenyl)methanamine | |
87736 | |
96% | |
3, 4, 5-Trimethoxybenzylamine, 96% |
95.0 | |
Colorless to Yellow | |
>110°C | |
95% min. (GC) | |
C10H15NO3 | |
(CH3O)3C6H2CH2NH2 | |
5 g | |
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine | |
COC1=CC(=CC(=C1OC)OC)CN | |
197.24 | |
197.24 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 242-468-0
TSCA : TSCA
RUO – Research Use Only